4-(Chloromethyl)-1H-imidazole hydrochloride 95%
Supplier: Ambeed
|
Danger
|
A100134-10G
A100134-1G
A100134-250MG
A100134-25G
A100134-5G
76982-104EA
202.64
USD
76982-104
76982-106
76982-108
76982-110
76982-112
4-(Chloromethyl)-1H-imidazole hydrochloride 95%
4-(Chloromethyl)-1H-imidazole hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₅ClN₂·ClH MW: 153.01 g/mol |
MDL Number:
MFCD01721661 CAS Number: 38585-61-4 UN: 1759 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...