Methyl 2-amino-5-chlorothiophene-3-carboxylate 95%

Supplier: Ambeed
Warning

A100131-100MG A100131-250MG A100131-1G A100131-5G
77008-256EA 39 USD
77008-256 77008-258 77889-694 77889-695
Methyl 2-amino-5-chlorothiophene-3-carboxylate 95%
Methyl 2-amino-5-chlorothiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MDL Number: MFCD20696783
CAS Number: 55503-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR