Boc-D-Lys(2-Cl-Z)-OH 98%

Supplier: Ambeed
Warning

A662479-1G A662479-25G A662479-5G
76980-866EA 14 USD
76980-866 76980-868 76980-870
Boc-D-Lys(2-Cl-Z)-OH 98%
Boc-D-Lys(2-Cl-Z)-OH

Precaución: for Research Use Only

Formula: C₁₉H₂₇ClN₂O₆
MDL Number: MFCD00065585
CAS Number: 57096-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR