2'-Amino-2'-deoxyuridine 98%

Supplier: Ambeed
Warning

A756305-100MG A756305-1G A756305-250MG
76981-404EA 24 USD
76981-404 76981-406 76981-408
2'-Amino-2'-deoxyuridine 98%
2'-Amino-2'-deoxyuridine

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O₅
MDL Number: MFCD01317284
CAS Number: 26889-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR