5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-((2R,3R,4R,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3-fluoro-4-hydroxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 2'-Deoxy-5'-O-DMT-2'-fluorouridine
A713577-100MG
A713577-1G
A713577-1MG
A713577-5G
A713577-25G
76981-172EA
10.61
USD
76981-172
76981-174
76981-176
76981-178
77673-958
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine 96%
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine
Precaución: for Research Use Only
Formula:
C₃₀H₂₉N₂O₇F MW: 548.56 g/mol |
MDL Number:
MFCD06657648 CAS Number: 146954-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...