1-((2R,3R,4R,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxy-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

Synonyms: 5’-O-(4,4’-Dimethoxytrityl)-2’-O-methyluridine

A313831-100MG A313831-10G A313831-1G A313831-250MG A313831-5G
76977-946EA 6.37 USD
76977-946 76977-948 76977-950 76977-952 77571-878
1-((2R,3R,4R,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxy-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione 95%
1-((2R,3R,4R,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxy-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₃₁H₃₂N₂O₈
MW: 560.59 g/mol
MDL Number: MFCD00274123
CAS Number: 103285-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR