(2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2'-OMe-A(Bz) Phosphoramidite
A293556-1G
A293556-5G
A293556-250MG
A293556-25G
76977-618EA
35
USD
76977-618
77612-692
77672-494
77881-806
(2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 97%
(2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
Precaución: for Research Use Only
Formula:
C₄₈H₅₄N₇O₈P MW: 887.95 g/mol |
MDL Number:
MFCD00792622 CAS Number: 110782-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...