(2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 97%
Supplier: Ambeed
|
Warning
|
A644597-10G
A644597-1G
A644597-25G
A644597-5G
A644597-250MG
A644597-100MG
76980-752EA
153
USD
76980-752
76980-754
76980-756
76980-758
77613-998
77673-750
(2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 97%
(2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
Precaución: for Research Use Only
Formula:
C₅₃H₆₆N₇O₈PSi |
MDL Number:
MFCD00080297 CAS Number: 104992-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...