(E)-Ethyl 3-(dimethylamino)acrylate 95%

Supplier: Ambeed
Warning

A255710-100G A255710-10G A255710-25G A255710-500G A255710-5G
76976-804EA 15 USD
76976-804 76976-806 76976-808 76976-810 76976-812
(E)-Ethyl 3-(dimethylamino)acrylate 95%
(E)-Ethyl 3-(dimethylamino)acrylate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD00144269
CAS Number: 1117-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR