Combretastatin A4 Phosphate disodium salt 98%

Supplier: Ambeed
Warning

A116116-10MG A116116-1MG A116116-250MG A116116-50MG A116116-5MG A116116-1G A116116-100MG
76971-176EA 76 USD
76971-176 76971-178 76971-180 76971-182 76971-184 77599-674 77610-946
Combretastatin A4 Phosphate disodium salt 98%
Combretastatin A4 Phosphate disodium salt

Precaución: for Research Use Only

Formula: C₁₈H₁₉Na₂O₈P
MDL Number: MFCD00943911
CAS Number: 168555-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR