N-Boc-S-tert-butyl-L-cysteine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Cys(tBu)-OH, (R)-2-((tert-Butoxycarbonyl)amino)-3-(tert-butylthio)propanoic acid

A184438-10G A184438-1G A184438-25G A184438-5G A184438-100G
76974-552EA 19.1 USD
76974-552 76974-554 76974-556 76974-558 77671-608
N-Boc-S-tert-butyl-L-cysteine 97%
N-Boc-S-tert-butyl-L-cysteine

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄S
MW: 277.379 g/mol
MDL Number: MFCD00076918
CAS Number: 56976-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR