2-Bromo-3-fluoroisonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-fluoropyridine-4-carboxylic acid

A330855-10G A330855-1G A330855-250MG A330855-25G A330855-5G
76978-238EA 257.8 USD
76978-238 76978-240 76978-242 76978-244 76978-246
2-Bromo-3-fluoroisonicotinic acid 97%
2-Bromo-3-fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 219.997 g/mol
MDL Number: MFCD13185796
CAS Number: 1211530-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR