2-Bromo-3-fluorobenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(bromomethyl)-3-fluorobenzene

A105452-1G A105452-25G A105452-250MG
76970-394EA 26 USD
76970-394 76970-396 77880-856
2-Bromo-3-fluorobenzyl bromide 97%
2-Bromo-3-fluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MDL Number: MFCD12911576
CAS Number: 1184918-22-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR