2-Cyano-4-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A384803-1G A384803-250MG
76978-864EA 82 USD
76978-864 76978-866
2-Cyano-4-fluorobenzoic acid 98%
2-Cyano-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄FNO₂
MDL Number: MFCD11110402
CAS Number: 1214369-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR