Boc-Ser(Tos)-OMe 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Methyl-2-((tert-butoxycarbonyl)amino)-3-(tosyloxy)propanoate, Boc-Ser(Tos)-OCH3, (S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(tosyloxy)propanoate
A373823-10G
A373823-1G
A373823-250MG
A373823-5G
76978-728EA
61.54
USD
76978-728
76978-730
76978-732
76978-734
Boc-Ser(Tos)-OMe 97%
Boc-Ser(Tos)-OMe
Precaución: for Research Use Only
Formula:
C₁₆H₂₃NO₇S MW: 373.42 g/mol |
MDL Number:
MFCD01632224 CAS Number: 56926-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...