1,5-Difluoro-3-methyl-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A172675-100MG A172675-250MG A172675-5G
76974-084EA 30.77 USD
76974-084 76974-086 76974-088
1,5-Difluoro-3-methyl-2-nitrobenzene 98%
1,5-Difluoro-3-methyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.117 g/mol
MDL Number: MFCD21603715
CAS Number: 1616526-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR