3-Chloro-4-fluoro-6-nitrotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-fluoro-5-methyl-4-nitrobenzene

A958934-1G A958934-5G
76982-018EA 33 USD
76982-018 76982-020
3-Chloro-4-fluoro-6-nitrotoluene 97%
3-Chloro-4-fluoro-6-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
MDL Number: MFCD21603714
CAS Number: 1352457-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR