1-Bromo-2,3-difluoro-4-methylbenzene 98%

Supplier: Ambeed
Warning

A484174-1G A484174-250MG A484174-5G
76979-762EA 34 USD
76979-762 76979-764 76979-766
1-Bromo-2,3-difluoro-4-methylbenzene 98%
1-Bromo-2,3-difluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MDL Number: MFCD21603711
CAS Number: 928304-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR