3-(Cyanomethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A214301-250MG A214301-1G
76975-618EA 21 USD
76975-618 77881-521
3-(Cyanomethyl)benzoic acid 97%
3-(Cyanomethyl)benzoic acid

Precaución: for Research Use Only

Formula: C9H7NO2
MDL Number: MFCD08234486
CAS Number: 5689-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR