2,3-Difluoro-1-methyl-4-nitrobenzene 98%

Supplier: Ambeed
Warning

A230237-10G A230237-1G A230237-5G A230237-25G
76976-122EA 55 USD
76976-122 76976-124 76976-126 77881-595
2,3-Difluoro-1-methyl-4-nitrobenzene 98%
2,3-Difluoro-1-methyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MDL Number: MFCD21603708
CAS Number: 932373-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR