2-Bromo-6-chlorobenzyl bromide 95%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-2-(bromomethyl)-3-chlorobenzene, 2-Chloro-α,6-dibromotoluene

A311882-1G A311882-250MG A311882-10G A311882-5G
76977-926EA 33.95 USD
76977-926 76977-928 77571-870 77672-592
2-Bromo-6-chlorobenzyl bromide 95%
2-Bromo-6-chlorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅Br₂Cl
MW: 284.376 g/mol
MDL Number: MFCD04038394
CAS Number: 75002-98-1
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR