2-(2-Chloroethyl)-1-methylpyrrolidine hydrochloride ≥95%

Supplier: Ambeed
Warning

A351322-1G A351322-5G
76978-512EA 66 USD
76978-512 76978-514
2-(2-Chloroethyl)-1-methylpyrrolidine hydrochloride ≥95%
2-(2-Chloroethyl)-1-methylpyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅Cl₂N
MW: 184.10 g/mol
MDL Number: MFCD00012717
CAS Number: 56824-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR