4'-Bromo-3'-chloro-2'-methylacetanilide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(4-Bromo-3-chloro-2-methylphenyl)acetamide
A209344-1G
A209344-5G
76975-484EA
21.22
USD
76975-484
76975-486
4'-Bromo-3'-chloro-2'-methylacetanilide 98%
4'-Bromo-3'-chloro-2'-methylacetanilide
Precaución: for Research Use Only
Formula:
C₉H₉BrClNO MW: 262.5300 g/mol |
MDL Number:
MFCD08059123 CAS Number: 125328-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...