Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 98%

Supplier: Ambeed
Warning

Synonyms: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl

A190393-100G A190393-1G A190393-25G A190393-500G A190393-5G A190393-1000G
76974-826EA 15 USD
76974-826 76974-828 76974-830 76974-832 77567-776 77611-834
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 98%
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)

Precaución: For research use only.

Formula: C₃H₈ClNO₂
MDL Number: MFCD00012870
CAS Number: 5680-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR