2-Chloro-4-fluoro-5-nitrotoluene 97%
Supplier: Ambeed
|
Warning
|
A244048-100G
A244048-10G
A244048-25G
A244048-500G
76976-546EA
39
USD
76976-546
76976-548
76976-550
76976-552
76976-554
2-Chloro-4-fluoro-5-nitrotoluene 97%
2-Chloro-4-fluoro-5-nitrotoluene
Precaución: for Research Use Only
Formula:
C₇H₅ClFNO₂ |
MDL Number:
MFCD11110549 CAS Number: 112108-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...