4-Bromo-3-fluorophenylacetonitrile 98%

Supplier: Ambeed
Warning

A571398-10G A571398-1G A571398-250MG A571398-25G A571398-5G
76980-326EA 110.34 USD
76980-326 76980-328 76980-330 76980-332 76980-334
4-Bromo-3-fluorophenylacetonitrile 98%
4-Bromo-3-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.0300 g/mol
MDL Number: MFCD12756199
CAS Number: 499983-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR