6-Amino-5-chloro-4-pyrimidinecarboxylic acid 98%

Supplier: Ambeed
Warning

A319939-100MG A319939-250MG
76978-054EA 5.31 USD
76978-054 76978-056
6-Amino-5-chloro-4-pyrimidinecarboxylic acid 98%
6-Amino-5-chloro-4-pyrimidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.56 g/mol
MDL Number: MFCD12923334
CAS Number: 914916-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR