Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-aminobicyclo[2.2.2]octane-1-carboxylate hydrochloride, 4-Aminobicyclo[2.2.2]octane-1-carboxylic acid methyl ester

A314396-100MG A314396-10G A314396-1G A314396-250MG A314396-5G
76977-960EA 56.23 USD
76977-960 76977-962 76977-964 76977-966 76977-968
Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate 95%
Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₂
MW: 183.2475 g/mol
MDL Number: MFCD20693756
CAS Number: 135908-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR