2-Biphenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-([1,1'-Biphenyl]-2-yl)acetic acid

A216968-100MG A216968-1G A216968-250MG
76975-726EA 42.44 USD
76975-726 76975-728 76975-730
2-Biphenylacetic acid 98%
2-Biphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
MDL Number: MFCD06208460
CAS Number: 14676-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR