1,12-Dodecanediol 97%

Supplier: Ambeed
Warning

A369292-1000G A369292-100G A369292-10G A369292-500G
76978-674EA 74 USD
76978-674 76978-676 76978-678 76978-680 76978-682
1,12-Dodecanediol 97%
1,12-Dodecanediol

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₂
MDL Number: MFCD00004755
CAS Number: 5675-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR