5-Mercapto-1H-1,2,3-triazole sodium salt 95%

Supplier: Ambeed
Danger

Synonyms: 1H-1,2,3-Triazole-5-thiol Sodium Salt, Sodium 1,2,3-triazole-5-thiolate

A462218-1G A462218-25G A462218-5G
76979-558EA 10 USD
76979-558 76979-560 76979-562
5-Mercapto-1H-1,2,3-triazole sodium salt 95%
5-Mercapto-1H-1,2,3-triazole sodium salt

Precaución: for Research Use Only

Formula: C₂H₃N₃S
MDL Number: MFCD00044799
CAS Number: 59032-27-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR