6-tert-Butylnicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-(tert-Butyl)nicotinic acid

A259255-100MG A259255-1G A259255-250MG
76976-880EA 23.34 USD
76976-880 76976-882 76976-884
6-tert-Butylnicotinic acid 95%
6-tert-Butylnicotinic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD08059362
CAS Number: 832715-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR