Tetra(3,5-dibromo-4-hydroxyphenyl)porphyrin 95%

Supplier: Ambeed
Warning

A858438-100MG A858438-1G A858438-250MG
76981-820EA 28 USD
76981-820 76981-822 76981-824
Tetra(3,5-dibromo-4-hydroxyphenyl)porphyrin 95%
Tetra(3,5-dibromo-4-hydroxyphenyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₄H₂₂Br₈N₄O₄
MDL Number: MFCD21608645
CAS Number: 125299-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR