α,4-Dibromo-3-fluorotoluene 97%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-3-fluorobenzyl bromide, 1-Bromo-4-(bromomethyl)-2-fluorobenzene

A761143-10G A761143-25G A761143-5G A761143-1G
76981-420EA 32 USD
76981-420 76981-422 76981-424 77882-653
α,4-Dibromo-3-fluorotoluene 97%
α,4-Dibromo-3-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MDL Number: MFCD11847179
CAS Number: 127425-73-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR