5-Amino-1-methyl-1H-pyrazole-4-carbonitrile 98%

Supplier: Ambeed
Danger

A146856-100G A146856-1G A146856-5G A146856-25G A146856-10G
76972-872EA 422.24 USD
76972-872 76972-874 76972-876 77592-748 77611-358
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile 98%
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₆N₄
MDL Number: MFCD00114569
CAS Number: 5334-41-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR