2-(4-Bromo-2,3,6-trifluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A475723-1G A475723-250MG
76979-696EA 102.91 USD
76979-696 76979-698
2-(4-Bromo-2,3,6-trifluorophenyl)acetic acid 98%
2-(4-Bromo-2,3,6-trifluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MW: 269.01 g/mol
MDL Number: MFCD03094472
CAS Number: 537033-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR