3-Fluoro-5-methylphenylacetonitrile 95%

Supplier: Ambeed
Danger

A558792-1G A558792-250MG
76980-260EA 58 USD
76980-260 76980-262
3-Fluoro-5-methylphenylacetonitrile 95%
3-Fluoro-5-methylphenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₈FN
MDL Number: MFCD03094317
CAS Number: 518070-21-8
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR