tert-Butyl (4-aminocyclohexyl)carbamate 97%

Supplier: Ambeed
Danger

Synonyms: N-Boc-1,4-diaminocyclohexane

A388427-100G A388427-10G A388427-1G A388427-250MG A388427-25G A388427-5G
76978-910EA 567 USD
76978-910 76978-912 76978-914 76978-916 76978-918 76978-920
tert-Butyl (4-aminocyclohexyl)carbamate 97%
tert-Butyl (4-aminocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD01076211
CAS Number: 195314-59-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR