2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)acetic acid hydrate 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Sar-OH·H₂O

A242282-100G A242282-25G A242282-5G
76976-502EA 50 USD
76976-502 76976-504 76976-506
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)acetic acid hydrate 98%
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)acetic acid hydrate

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₅
MDL Number: MFCD00180724
CAS Number: 212651-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR