4-Isothiocyanato-2-(trifluoromethyl)benzonitrile 98%
Supplier: Ambeed
|
Danger
|
A597381-5G
A597381-25G
76980-484EA
12
USD
76980-484
77882-408
4-Isothiocyanato-2-(trifluoromethyl)benzonitrile 98%
4-Isothiocyanato-2-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₉H₃F₃N₂S |
MDL Number:
MFCD09800709 CAS Number: 143782-23-4 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...