7-MAC 98%

Supplier: Ambeed
Danger

Synonyms: Diphenylmethyl (6R,7S)-7-Amino-7-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Diphenylmethyl 7beta-Amino-7alpha-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-3-cephem-4-carboxylate, Benzhydryl (6R,7S)-7-amino-7-methoxy-3-(((1-methyl-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

A179369-100G A179369-10G A179369-1G A179369-25G A179369-5G
76974-392EA 162.32 USD
76974-392 76974-394 76974-396 76974-398 76974-400
7-MAC 98%
7-MAC

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₆O₄S₂
MW: 524.6152 g/mol
MDL Number: MFCD08704705
CAS Number: 56610-72-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR