4-Amino-3,6-dichloropyridazine 97%

Supplier: Ambeed
Warning

A144660-100G A144660-10G A144660-1G A144660-25G A144660-5G
76972-754EA 401.03 USD
76972-754 76972-756 76972-758 76972-760 76972-762
4-Amino-3,6-dichloropyridazine 97%
4-Amino-3,6-dichloropyridazine

Precaución: for Research Use Only

Formula: C₄H₃Cl₂N₃
MW: 163.9900 g/mol
MDL Number: MFCD01647253
CAS Number: 823-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR