4-Bromo-3-iodobenzoic acid 97%

Supplier: Ambeed
Warning

A122145-10G A122145-1G A122145-5G
76971-598EA 48.81 USD
76971-598 76971-600 76971-602
4-Bromo-3-iodobenzoic acid 97%
4-Bromo-3-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrIO₂
MW: 326.914 g/mol
MDL Number: MFCD12406813
CAS Number: 42860-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR