1-(tert-Butoxycarbonyl)-3-iodopyrrolidine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-iodo-pyrrolidine, tert-Butyl 3-iodopyrrolidine-1-carboxylate

A286162-10G A286162-1G A286162-250MG A286162-25G A286162-100MG A286162-5G
76977-512EA 147 USD
76977-512 76977-514 76977-516 77672-478 77881-785 77881-786
1-(tert-Butoxycarbonyl)-3-iodopyrrolidine 95%
1-(tert-Butoxycarbonyl)-3-iodopyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₆INO₂
MDL Number: MFCD12031283
CAS Number: 774234-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR