Methyl-3-chloro-4-methylbenzoate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-chloro-4-methylbenzoate

A488992-100G A488992-25G A488992-5G
76979-778EA 105 USD
76979-778 76979-780 76979-782
Methyl-3-chloro-4-methylbenzoate 96%
Methyl 3-chloro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD00205141
CAS Number: 56525-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR