3-Amino-1-benzhydrylazetidine mesylate 95%

Supplier: Ambeed
Warning

A136753-1G A136753-5G
76972-362EA 52 USD
76972-362 76972-364
3-Amino-1-benzhydrylazetidine mesylate 95%
3-Amino-1-benzhydrylazetidine mesylate

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₃S
MDL Number: MFCD20489148
CAS Number: 1373253-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR