5-Oxo-L-proline L-arginine salt 98%

Supplier: Ambeed
Warning

A460636-1000G A460636-100G A460636-25G A460636-500G A460636-5G
76979-528EA 57 USD
76979-528 76979-530 76979-532 76979-534 76979-536
5-Oxo-L-proline L-arginine salt 98%
5-Oxo-L-proline L-arginine salt

Precaución: for Research Use Only

Formula: C₁₁H₂₁N₅O₅
MDL Number: MFCD17976754
CAS Number: 56265-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR