Sucralose 98%

Supplier: Ambeed
Warning

A551773-100G A551773-25G A551773-5G
76980-232EA 51.01 USD
76980-232 76980-234 76980-236
Sucralose 98%
Sucralose

Precaución: for Research Use Only

Formula: C₁₂H₁₉Cl₃O₈
MDL Number: MFCD03648615
CAS Number: 56038-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR