Benzyl 3-aminopyrrolidine-1-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A854353-1G A854353-250MG A854353-5G
76981-808EA 24 USD
76981-808 76981-810 76981-812
Benzyl 3-aminopyrrolidine-1-carboxylate hydrochloride 95%
Benzyl 3-aminopyrrolidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClN₂O₂
MW: 256.73 g/mol
MDL Number: MFCD09955427
CAS Number: 1159822-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR