2-Methoxy-6-methyl-4(1H)-pyrimidinone 97%

Supplier: Ambeed
Warning

A104937-100MG A104937-1G A104937-250MG A104937-5G
76970-364EA 6 USD
76970-364 76970-366 76970-368 77880-848
2-Methoxy-6-methyl-4(1H)-pyrimidinone 97%
2-Methoxy-6-methyl-4(1H)-pyrimidinone

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD09743870
CAS Number: 55996-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR